Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DAPI
GREENER_CHOICESDP

Catalog No. 574810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-481-0 10 mg
57-485-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-481-0 Supplier Tocris Bioscience™ Supplier No. 5748/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fluorescent DNA stain

DAPI is a fluorescent DNA stain/ dye. DAPI stain binds to AT-rich regions of DNA and fluorescence increases by approximately 20-fold when bound to double-stranded DNA. DAPI stain can be used in flow cytometry, DNA visualization and quantification in histochemistry/biochemistry, to assess apoptosis, and to visualize mouse embryos or fetal organs. DAPI stain can be used in fixed cells and is cell-permeable at high concentrations providing effective cell staining. It is reported to be more photostable than Hoechst 33342 (Cat. No. 5117). Em/Ex λ = 350/470 nm respectively.

Specifications

Chemical Name or Material 2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride
CAS 28718-90-3
Quantity 10 mg
Target Fluorescent DNA Stains
Molecular Formula C16H15N50.2 HCl
Formula Weight 350.25
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.