Learn More
Promega DAPK1 Kinase Enzyme System, 1mg
Catalog No. PRV6390 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6390 1 mg
1 options
Catalog No. PRV6390 Supplier Promega Supplier No. V6390
Add to Cart
Add to Cart
Request a Quote
Recombinant human DAPK1 (1-363) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Death-associated protein kinase 1 (DAPK1) is a positive mediator of apoptosis induced by gamma-interferon.
  • Easily Screen and Profile DAPK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C