Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAPK1 Monoclonal Antibody (12B1)
GREENER_CHOICE

Catalog No. PIMA536128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536128 Supplier Invitrogen™ Supplier No. MA536128
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAPK1 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

DAPK1 is a large, calmodulin-dependent kinase involved in gamma-interferon induced programmed cell death and is a potential tumor suppressor. DAPK1 is involved in multiple cellular signaling pathways that trigger cell survival, apoptosis, and authophagy. It regulates both type I apoptotic and type II autophagic cell death signal, depending on the cellular setting. Type I apoptotic cell death is caspase-dependent, type II authophagic cell death is caspase-independent. DAPK1 phosphorylates PIN1 resulting in inhibition of its catalytic activity, nuclear localization, and cellular function. It also phosphorylates TPM1, STX1A, PRKD1, BECN1, TSC2, RPS6, MYL9, and DAPK3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAPK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 12B1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene DAPK1
Gene Accession No. P53355, Q80YE7
Gene Alias D13Ucla1; DAP kinase 1; DAPK; DAPK1; DAP-Kinase; death associated protein kinase 1; death-associated protein kinase 1; DKFZp781I035; OTTHUMP00000216193
Gene Symbols DAPK1
Host Species Mouse
Immunogen Recombinant protein within Human DAPK1 aa 447-683.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1612, 306722, 69635
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.