Learn More
Invitrogen™ DAPK2 Polyclonal Antibody

Description
This antibody will not cross-react with DAPK1. In Western blot applications, this antibody detects a band at ~42kDa. A suggested positive control is A431 cell lysate. PA5-19961 can be used with blocking peptide PEP-0086.
Specifications
Specifications
| Antigen | DAPK2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | DAPK2 |
| Gene Accession No. | Q8VDF3, Q9UIK4 |
| Gene Alias | DAP kinase 2; DAPK2; DAP-kinase-related protein 1; DAP-kinase-related protein 1 beta isoform; death associated protein kinase 2; death-associated kinase 2; death-associated protein kinase 2; DRAK2; DRP1; DRP-1; MGC119312 |
| Gene Symbols | DAPK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.