Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAPK3/ZIPK Rabbit anti-Human, Clone: S02-7F5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315020100UL
This item is not returnable.
View return policy
Description
DAPK3/ZIPK Monoclonal antibody specifically detects DAPK3/ZIPK in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
DAPK3/ZIPK | |
Monoclonal | |
Unconjugated | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
S02-7F5 | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
DAP kinase 3, DAP-like kinase, death-associated protein kinase 3, dlk, EC 2.7.11, EC 2.7.11.1, ZIP, ZIP kinase isoform, ZIPKFLJ36473, ZIP-kinase | |
A synthetic peptide of human DAPK3/ZIPK (Uniprot#: O43293) | |
100 μg | |
Apoptosis | |
1613 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction