Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549251
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549251 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549251 Supplier Invitrogen™ Supplier No. PA549251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAPL1 Polyclonal Antibody for Western Blot

May play a role in the early stages of epithelial differentiation or in apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAPL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DAPL1
Gene Accession No. A0PJW8, Q9D757
Gene Alias 2310032F03Rik; Dapl1; death associated protein like 1; death associated protein-like 1; death-associated protein-like 1; Early epithelial differentiation-associated protein; EEDA; RGD1309610
Gene Symbols DAPL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 20-48 amino acids from the N-terminal region of human DAPL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76747, 92196
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.