Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAPP1 Monoclonal Antibody (4H9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531546
Description
Keep as concentrated solution. Positive Control: TETN Ramos. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
Specifications
DAPP1 | |
Monoclonal | |
2.36 mg/mL | |
PBS with no preservative | |
Q9UN19 | |
DAPP1 | |
The immunogen used to generate this antibody corresponds to human DAPP1. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
4H9 | |
Unconjugated | |
DAPP1 | |
b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1 | |
Mouse | |
Protein A | |
RUO | |
27071, 362046 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction