Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAPP1 Mouse anti-Human, Clone:2F7A9, Abnova™

Catalog No. 89333653 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-653 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-653 Supplier Abnova Corporation Supplier No. MAB17230
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human DAPP1.

Specifications

Antigen DAPP1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2F7A9
Conjugate Unconjugated
Description dual adaptor of phosphotyrosine and 3-phosphoinositides
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene DAPP1
Gene Alias BAM32, DKFZp667E0716
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 127-276 of human DAPP1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 27071
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.