Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAPP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528575
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Raji. Predicted reactivity: Mouse (93%), Rat (91%), Chicken (86%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
Specifications
| DAPP1 | |
| Polyclonal | |
| Unconjugated | |
| DAPP1 | |
| b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UN19 | |
| DAPP1 | |
| Recombinant protein encompassing a sequence within the center region of human DAPP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction