Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAPP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982517
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2517 Supplier Bioss Supplier No. BS12982R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.

Specifications

Antigen DAPP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DAPP1
Gene Accession No. Q9UN19
Gene Alias B lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1
Gene Symbols DAPP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DAPP1/BAM32.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27071
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.