Learn More
DAPT
Description
DAPT is used in the study of β-amyloid formation. DAPT has been shown to inhibit Notch signaling in studies of autoimmune and lymphoproliferative diseases, such as ALPS and lupus erythematosus, as well as in cancer cell growth, angiogenesis, and differentiation of human induced pluripotent stem cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
DAPT is used in the study of β-amyloid formation. DAPT has been shown to inhibit Notch signaling in studies of autoimmune and lymphoproliferative diseases, such as ALPS and lupus erythematosus, as well as in cancer cell growth,
Specifications
Specifications
| Color | White |
| Quantity | 10 mg |
| Sensitivity | Light Sensitive |
| Solubility Information | Soluble in DMSO,ethanol,dichloromethane,DMF. |
| Formula Weight | 432.5 |
| Physical Form | Lyophilized |
| Chemical Name or Material | DAPT |
Safety and Handling
| TSCA | No |
| Recommended Storage | Store at -20°C; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.