Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ DAPT
GSA_VAGREENER_CHOICESDP

Catalog No. 263450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-345-0 50 mg
26-341-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-345-0 Supplier R&D Systems™ Supplier No. 2634/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Gamma γ-secretase inhibitor; induces neuronal differentiation; blocks Notch signaling

DAPT is a γ-secretase inhibitor. DAPT reduces Aβ40 and Aβ42 levels in human primary neuronal cultures (IC50 values are 115 and 200 nM for total Aβ and Aβ42 respectively) and in brain extract, cerebrospinal fluid and plasma in vivo. DAPT has no effect on APPα and APPβ levels. DAPT blocks Notch signaling in hybrid human-mouse fetal thymus organ culture (FTOC) and causes ESCs to commit to neuronal differentiation. DAPT can be used in a small molecule cocktail to derive cortical neurons from hPSCs and to maintain hepatocytes in culture. DAPT also promotes the formation of cone photoreceptors in retinal organoids.

For more information about how DAPT may be used, see our protocols: Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts, Generating Midbrain Dopaminergic Neurons from hPSCs .

DAPT synthesized to Ancillary Material Grade also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 208255-80-5
Molecular Formula C23H26F2N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 432.46
Purity >99%
Molecular Weight (g/mol) 432.468
Chemical Name or Material DAPT
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.