Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAPT, PROMO, Thermo Scientific Chemicals

Catalog No. J65864MAPM
Click to view available options
Quantity:
10 mg

Applications

DAPT is used in the study of B-amyloid formation. DAPT has been shown to inhibit Notch signaling in studies of autoimmune and lymphoproliferative diseases, such as ALPS and lupus erythematosus, as well as in cancer cell growth,

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208255-80-5
Molecular Formula C23H26F2N2O4
Molecular Weight (g/mol) 432.47 g/mol
MDL Number MFCD04974585
InChI Key DWJXYEABWRJFSP-XOBRGWDASA-N
Synonym dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
PubChem CID 5311272
ChEBI CHEBI:86193
IUPAC Name tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate
SMILES CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F

Specifications

Color White
Quantity 10 mg
Sensitivity Light Sensitive
Solubility Information Soluble in DMSO, ethanol, dichloromethane, DMF.
Physical Form Lyophilized Powder
Chemical Name or Material DAPT
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.