Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARC Antibody (18B11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DARC |
---|---|
Clone | 18B11 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
DARC Monoclonal antibody specifically detects DARC in Human samples. It is validated for Flow Cytometry, ELISASpecifications
DARC | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
A synthesized peptide derived from Human DARC [UniProt Q16570] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
18B11 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
GPCR, Immunology, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2532 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title