Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARC Antibody (2C3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275196
Description
DARC Monoclonal specifically detects DARC in Human samples. It is validated for Western Blot, ELISA, Flow (Intracellular), Immunofluorescence, Block/Neutralize.Specifications
DARC | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
ACKR1 | |
CHO cells transfected with cDNA encoding the Duffy glycoprotein. | |
0.2 mg | |
2532.0 | |
Human | |
Purified |
Neutralization, ELISA, Flow Cytometry, Immunofluorescence, Blocking Assay, Neutralization | |
2C3 | |
Western Blot, ELISA 1:100 - 1:2000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence, Block/Neutralize | |
CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
Mouse | |
Protein A purified | |
Primary | |
The antibody binds specifcally to the pentapeptide FEDVW of the Duffy receptor. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction