Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARC Antibody (358307), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4139M
Description
DARC Monoclonal antibody specifically detects DARC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
DARC | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
358307 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
HEK293 human embryonic kidney cell line transfected with human DARC, Met1-Ser336, Accession # Q16570 | |
0.1 mL | |
GPCR, Immunology, Innate Immunity | |
2532 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction