Learn More
Invitrogen™ DARC Polyclonal Antibody

Description
A suggested positive control is PC-3 cell lysate. PA5-20360 can be used with blocking peptide PEP-0585.
Specifications
Specifications
| Antigen | DARC |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ACKR1 |
| Gene Accession No. | Q16570 |
| Gene Alias | AA162249; ACKR1; Atypical chemokine receptor 1; atypical chemokine receptor 1 (Duffy blood group); CCBP1; CD234; CD234 antigen; CTA-134P22.3; DARC; Dfy; Duffy Antigen; Duffy antigen chemokine receptor; Duffy antigen receptor for chemokines; duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; ESTM35; FY; Fy Antigen; Fy glycoprotein; FYDuffy blood group; glycoprotein D; GPD; GPDWBCQ1; GpFy; HGNC:4035; OTTHUMP00000060081; Plasmodium vivax receptor; WBCQ1 |
| Gene Symbols | ACKR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.