Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DARC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582549 Supplier Invitrogen™ Supplier No. PA582549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CHQATRTLLP SLPLPEGWSS HLDTLGSKS Highest antigen sequence indentity to the following orthologs: Rat - 38%, Mouse - 42%.

ACKR1 or DARC is an atypical chemokine receptor that controls chemokine levels and localization via high-affinity chemokine binding that is uncoupled from classic ligand-driven signal transduction cascades resulting instead in chemokine sequestration, degradation, or transcytosis. ACKR1 is also known as an interceptor (internalizing receptor) or chemokine-scavenging receptor or chemokine decoy receptor. It has a promiscuous chemokine-binding profile by interacting with inflammatory chemokines of both the CXC and the CC subfamilies but not with homeostatic chemokines. ACKR1 also acts as a receptor for chemokines including CCL2, CCL5, CCL7, CCL11, CCL13, CCL14, CCL17, CXCL5, CXCL6, IL8/CXCL8, CXCL11, GRO, RANTES, MCP-1, TARC and for the malaria parasites P.vivax and P.knowlesi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DARC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACKR1
Gene Accession No. Q16570
Gene Alias AA162249; ACKR1; Atypical chemokine receptor 1; atypical chemokine receptor 1 (Duffy blood group); CCBP1; CD234; CD234 antigen; CTA-134P22.3; DARC; Dfy; Duffy Antigen; Duffy antigen chemokine receptor; Duffy antigen receptor for chemokines; duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; ESTM35; FY; Fy Antigen; Fy glycoprotein; FYDuffy blood group; glycoprotein D; GPD; GPDWBCQ1; GpFy; HGNC:4035; OTTHUMP00000060081; Plasmodium vivax receptor; WBCQ1
Gene Symbols ACKR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACKR1. Recombinant protein control fragment (Product #RP-90870).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.