Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARC Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP1772780.025MG
Description
DARC Polyclonal antibody specifically detects DARC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DARC | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q16570 | |
CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
GPCR, Immunology, Innate Immunity | |
Primary | |
2532 |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Affinity Purified | |
DARC | |
Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human DARC. The immunogen is located within the last 50 amino acids of DARC. | |
35 kDa | |
0.025 mg | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title