Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARPP-32 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB300304
Description
DARPP-32 Polyclonal specifically detects DARPP-32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DARPP-32 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250 | |
Q6J4I0 | |
PPP1R1B | |
Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # Q6J4I0 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼32k DARPP-32 protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32) | |
Rabbit | |
32 kDa | |
0.1 mL | |
Breast Cancer, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase, Signal Transduction | |
84152 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction