Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DARPP-32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596173 Supplier Invitrogen™ Supplier No. PA596173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPKDRKKIQ FSVPAPPSQL DPRQVEMIRR RRPTPAMLFR LSEHSSPEEE ASPHQRASGE GHHLKSKRPN PCAYTPPSLK AVQRIAESHL QSISNLNENQ ASEEEDELGE LRELGY Positive Samples: 293T; Cellular Location: Cytoplasm.

DARPP-32 is a dopamine (DA) and AMP-regugated ~32 kDa phosphoprotein that is associated with dopaminoceptive neurons. The protein inhibits Protein Phosphatase I when it is phosphorylated on Thr34. In contrast, when DARPP-32 is phosphorylated on Thr75 the protein acts as an inhibitor of PKA. Phosphorylation of DARPP-32 is thought to play a critical role in the regugation of dopaminergic neurotransmission. In addition, the activity of DARPP-32 is also thought to play important roles in the actions of alcohol, caffeine and Prozac™.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DARPP-32
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Ppp1r1b
Gene Accession No. Q60829, Q6J4I0, Q9UD71
Gene Alias AU040756; DARPP32; DARPP-32; dopamine- and cAMP-regulated neuronal phosphoprotein; dopamine and cAMP-regulated neuronal phosphoprotein 32; dopamine- and cAMP-regulated phosphoprotein DARPP-32; FLJ20940; IPPD; neuronal phosphoprotein DARPP-32; OTTHUMP00000164275; OTTHUMP00000164276; Ppp1r1b; PPR1B; protein pho; protein phosphatase 1 regulatory inhibitor subunit 1B; protein phosphatase 1 regulatory subunit 1B; protein phosphatase 1, regulatory (inhibitor) subunit 1B
Gene Symbols Ppp1r1b
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19049, 360616, 84152
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.