Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARS Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300268FR
Description
DARS Polyclonal specifically detects DARS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DARS | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
aspartate tRNA ligase 1, cytoplasmic, Aspartate--tRNA ligase, aspartyl-tRNA synthetase, aspartyl-tRNA synthetase, cytoplasmic, aspRS, Cell proliferation-inducing gene 40 protein, cell proliferation-inducing protein 40, DKFZp781B11202, EC 6.1.1.12, MGC111579 | |
Produced in rabbits immunized with purified, recombinant DARS (P14868; Met1-Pro501). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
1615 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction