Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DARS Monoclonal Antibody (11)
GREENER_CHOICE

Catalog No. PIMA529211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529211 Supplier Invitrogen™ Supplier No. MA529211
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DARS.

Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DARS
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DARS1
Gene Accession No. P14868
Gene Alias 5730439G15Rik; aspartate tRNA ligase 1, cytoplasmic; aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; aspRS; Cell proliferation-inducing gene 40 protein; Dars; DRS1; HBSL; PIG40; testicular tissue protein Li 192
Gene Symbols DARS1
Host Species Mouse
Immunogen Recombinant Human DARS Protein (Met1-Pro501).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1615
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.