Learn More
Invitrogen™ DARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527892
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (94%), Zebrafish (83%), Xenopus laevis (90%), Chicken (90%), Sheep (94%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
Specifications
DARS | |
Polyclonal | |
Unconjugated | |
DARS1 | |
5730439G15Rik; aspartate tRNA ligase 1, cytoplasmic; aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; aspRS; Cell proliferation-inducing gene 40 protein; Dars; DRS1; HBSL; PIG40; testicular tissue protein Li 192 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.35 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P14868 | |
DARS1 | |
Recombinant protein encompassing a sequence within the center region of human DARS. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.