Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556266 Supplier Invitrogen™ Supplier No. PA556266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPSASASRKS QEKPREIMDA AEDYAKERYG ISSMIQSQEK PDRVLVRVRD LTIQKADEVV WVRARVHTSR AKGKQCFLVL RQQQFNVQAL VAVGDHASKQ MVKFAANINK ESIVDVEGVV RKVNQKIGSC TQQDV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DARS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DARS1
Gene Accession No. P14868
Gene Alias 5730439G15Rik; aspartate tRNA ligase 1, cytoplasmic; aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; aspRS; Cell proliferation-inducing gene 40 protein; Dars; DRS1; HBSL; PIG40; testicular tissue protein Li 192
Gene Symbols DARS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DARS. Recombinant protein control fragment (Product #RP-92462).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1615
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.