Learn More
Fisherbrand™ Traceable™ Data Acquisition Software
Complete Data Acquisition Software enables measurement readings to be displayed and stored on a PC from up to 8 devices at a time.
$267.82 - $978.27
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
03-391-121
|
Fisherbrand™
3780413 |
Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections) |
Each for $267.82
|
|
|||||
03-391-124
|
Fisherbrand™
9008021 |
Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections) |
Each for $978.27
|
|
|||||
Description
- Display and store measurement readings on a PC from up to 8 devices at a time
- Data capture runs in the background, enabling users to work on other programs simultaneously
- Set audible and visual notification alarms, or immediately send an email if an over/under condition occurs
- Software can be used to connect a meter directly to PC via cable
- Check software specification tables for meter compatibility
Specifications
Data Acquisition Software | |
No Display |