Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DATF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101330 Supplier Invitrogen™ Supplier No. PA5101330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DIDO1.

Dio-1 (Death Inducer-Obliterator-1) is a putative transcription factor that contains two zinc finger motifs. Dio-1 translocates to the nucleus, and activates apoptosis during limb development. Programmed cell death, a highly regulated form of apoptosis, plays an important role in determining the amount of tissue, the shape, and the definition of each digit during limb development. Dio-1 expression is upregulated when an apoptotic signal is detected, and subsequently apoptosis is induced. This process is similar to the expression of NFkB and NGF in response to external signals. Dio-1 expression is suppressed by caspase inhibitors and Bcl-2 expression. This supports the theory that Dio-1 functions in the onset of programmed cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DATF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DIDO1
Gene Accession No. Q8C9B9, Q9BTC0
Gene Alias 6720461J16Rik; BYE1; C20orf158; D130048F08Rik; DATF1; DATF-1; death associated transcription factor 1; death inducer-obliterator 1; death inducer-obliterator-2; death inducer-obliterator-3; death-associated transcription factor 1; Death-inducer obliterator 1; dido; Dido1; DIDO2; DIDO3; DIO1; DIO-1; dJ885L7.8; KIAA0333
Gene Symbols DIDO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DIDO1(Accession Q9BTC0), corresponding to amino acid residues P2006-G2056.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11083, 23856
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.