Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daunorubicin hydrochloride

Catalog No. AAJ60224MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60224MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60224MA Supplier Thermo Scientific Chemicals Supplier No. J60224MA
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23541-50-6 | C27H30ClNO10 | 563.984 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23541-50-6
Molecular Formula C27H30ClNO10
Molecular Weight (g/mol) 563.984
MDL Number MFCD04974507
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
Synonym daunorubicin hydrochloride, daunorubicin hcl, daunomycin hydrochloride, daunoblastina, rubidomycin hydrochloride, daunorubicinol hydrochloride, daunoblastin, rubomycin, ondena, daunomycin chlorohydrate
PubChem CID 62770
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl

Specifications

Color Red
Quantity 10 mg
UN Number UN2811
Beilstein 4229221
Sensitivity Hygroscopic
Merck Index 14,2832
Formula Weight 563.98
Physical Form Powder
Chemical Name or Material Daunorubicin hydrochloride
Hazard Category H301-H317-H334-H351
Hazard Statement GHS H Statement
H301-H334-H351-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P310-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 245-723-4
RTECSNumber HB7878000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.