Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Daunorubicin hydrochloride
GREENER_CHOICESDP

Catalog No. 146710
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-671-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-671-0 Supplier Tocris Bioscience™ Supplier No. 1467/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DNA topoisomerase II inhibitor

Inhibits RNA and DNA synthesis and causes DNA fragmentation in vivo. Reduces tau mRNA levels in vitro.Anticancer agent. Exhibits activity nonlymphocytic leukemia. Identified as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 23541-50-6
Molecular Formula C27H30ClNO10

Specifications

Quantity 10 mg
Formula Weight 563.99
Purity >98%
Molecular Weight (g/mol) 563.984
Chemical Name or Material Daunorubicin hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.