Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Patient Belonging Bags

These Patient Belonging Bags are for holding patient's belongings and have a drawstring closure that can easily be hung from wheelchair or gurney.

$45.75

Specifications

Capacity (English) 5 lb.
Closure Type Drawstring Closure
Dimensions (L x W) 20 x 20 in.
Material Plastic
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-415-783
View Documents
Dukal™
DSPB01
White
Case of 250 for $45.75
Add to cart
 
22-415-784
View Documents
Dukal™
DSPB01C
Clear
Case of 250 for $45.75
Add to cart
 
Description

Description

  • Durable for holding patient's clothing and belongings.
  • Can easily be hung from wheelchair or gurney.
  • Designated space to write patient's name and room number.
Specifications

Specifications

5 lb.
Drawstring Closure
20 x 20 in.
Plastic
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit