Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Roll On Antiperspirant and Deodorant

Catalog No. 22415792
Change view
Click to view available options
Volume (English):
1.5 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-415-792 1.5 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-415-792 Supplier Dukal™ Supplier No. RD15
Add to Cart
Add to Cart

This Roll On Antiperspirant and Deodorant fights wetness and odor.

  • Fights wetness and odor
  • Fresh scent

Specifications

Product Type Roll-on Deodorant
Color Clear
For Use With (Application) Personal Care, Deodorant
Volume (English) 1.5 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.