Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Toothbrushes

Catalog No. 22415799
Click to view available options
No. of Tufts:
30 Tufts
52 Tufts
Color:
Blue with White Bristles
Ivory with Clear Bristles
This item is not returnable. View return policy

Catalog No. 22-415-799

Supplier: Dukal™ TB52

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

These Toothbrushes feature soft bristles with rounded polish tips.

  • Features soft bristles with rounded polish tips.
  • Individually packaged.

Specifications

Product Type Toothbrush
Color Blue with White Bristles
Material Plastic
For Use With (Application) Oral Care
For Use With (Equipment) Toothpaste
No. of Tufts 52 Tufts
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.