Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dukal™ DawnMist™ Toothbrushes
Click to view available options
No. of Tufts:
30 Tufts
52 Tufts
Color:
Blue with White Bristles
Ivory with Clear Bristles
Description
- Features soft bristles with rounded polish tips.
- Individually packaged.
Specifications
Specifications
| Product Type | Toothbrush |
| Color | Ivory with Clear Bristles |
| Material | Plastic |
| For Use With (Application) | Oral Care |
| For Use With (Equipment) | Toothpaste |
| No. of Tufts | 30 Tufts |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction