Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Daxx Antibody (3A2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Daxx |
---|---|
Clone | 3A2 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for ELISASpecifications
Daxx | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
A synthesized peptide derived from Human Daxx [UniProt Q9UER7] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3A2 | |
ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1616 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title