Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Daxx Antibody (PCRP-DAXX-5G11), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314025PE
This item is not returnable.
View return policy
Description
Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Flow CytometrySpecifications
Daxx | |
Monoclonal | |
PE | |
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
Recombinant full-length human Daxx protein (Uniprot: Q9UER7) | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
1616 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-DAXX-5G11 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction