Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Daxx Antibody (PCRP-DAXX-8C2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308741
Description
Daxx Monoclonal specifically detects Daxx in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Daxx | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry : 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Immunohistochemistry-Paraffin | |
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
Recombinant full-length human Daxx protein | |
100 μg | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
1616 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-DAXX-8C2 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction