Learn More
Invitrogen™ DAXX Monoclonal Antibody (DAXX-01)

Description
This antibody recognizes Daxx, a cytoplasmic death domain containing protein mainly expressed in fetal and adult human and mouse tissue.
Specifications
Specifications
| Antigen | DAXX |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | DAXX-01 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | DAXX |
| Gene Accession No. | Q9UER7 |
| Gene Alias | BING2; CENP-C binding protein; DADB-159G18.9; DAP6; Daxx; death domain associated protein; Death domain-associated protein 6; death-associated protein 6; death-domain associated protein; EAP1; ETS1-associated protein 1; Fas death domain-associated protein; Fas-binding protein; hDaxx; MGC126245; MGC126246; SMIM40; Unknown (protein for MGC:138055) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.