Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Daxx Rabbit anti-Human, Clone: 5B3X4, Novus Biologicals™

Rabbit Monoclonal Antibody
$196.25 - $452.15
Specifications
| Antigen | Daxx |
|---|---|
| Clone | 5B3X4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Western BlotSpecifications
| Daxx | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1616 | |
| IgG | |
| Affinity purified |
| 5B3X4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Daxx (Q9UER7). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title