Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAXX Recombinant Rabbit Monoclonal Antibody (5B3X4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537945
Description
Positive Samples: K-562, SH-SY5Y Immunogen sequence: AEIRRLQEKE LDLSELDDPD SAYLQEARLK RKLIRLFGRL CELKDCSSLT GRVIEQRIPY RGTRYPEVNR RIERLINKPG PDTFPDYGDV LRAVEKAAAR H.
This gene encodes a multifunctional protein that resides in multiple locations in the nucleus and in the cytoplasm. It interacts with a wide variety of proteins, such as apoptosis antigen Fas, centromere protein C, and transcription factor erythroblastosis virus E26 oncogene homolog 1. In the nucleus, the encoded protein functions as a potent transcription repressor that binds to sumoylated transcription factors. Its repression can be relieved by the sequestration of this protein into promyelocytic leukemia nuclear bodies or nucleoli. This protein also associates with centromeres in G2 phase. In the cytoplasm, the encoded protein may function to regulate apoptosis. The subcellular localization and function of this protein are modulated by post-translational modifications, including sumoylation, phosphorylation and polyubiquitination. Alternative splicing results in multiple transcript variants.
Specifications
DAXX | |
Recombinant Monoclonal | |
0.45 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UER7 | |
DAXX | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Daxx (Q9UER7). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
5B3X4 | |
Unconjugated | |
DAXX | |
BING2; CENP-C binding protein; DADB-159G18.9; DAP6; Daxx; death domain associated protein; Death domain-associated protein 6; death-associated protein 6; death-domain associated protein; EAP1; ETS1-associated protein 1; Fas death domain-associated protein; Fas-binding protein; hDaxx; MGC126245; MGC126246; SMIM40; Unknown (protein for MGC:138055) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1616 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction