Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAZ1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | DAZ1 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DAZ1 Polyclonal antibody specifically detects DAZ1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DAZ1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q86SG3 | |
1617 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2), 40% Glycerol | |
deleted in azoospermia, deleted in azoospermia 1, FLJ17175, SPGYDAZdeleted in azoospermia protein 1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: NDVDVQKIVGSQIHFHGKKLKLGPAIRKQ | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title