Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAZ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544927
Description
Peptide sequence: MSAANPETPN STISREASTQ SSSAAASQGW VLPEGKIVPN TVFVGGIDAR Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 85%; Rat: 79%.
This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor. Its expression is restricted to premeiotic germ cells, particularly in spermatogonia. It encodes an RNA-binding protein that is important for spermatogenesis. Four copies of this gene are found on chromosome Y within palindromic duplications; one pair of genes is part of the P2 palindrome and the second pair is part of the P1 palindrome. Each gene contains a 2.4 kb repeat including a 72-bp exon, called the DAZ repeat; the number of DAZ repeats is variable and there are several variations in the sequence of the DAZ repeat. Each copy of the gene also contains a 10.8 kb region that may be amplified; this region includes five exons that encode an RNA recognition motif domain. This gene contains three copies of the 10.8 kb repeat. However, no transcripts containing three copies of the RRM domain have been described; thus the RefSeq for this gene contains only two RRM domains.
Specifications
DAZ1 | |
Polyclonal | |
Unconjugated | |
DAZ1 | |
DAZ; DAZ1; deleted in azoospermia 1; deleted in azoospermia protein 1; SPGY; testicular tissue protein Li 49 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1617 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NQZ3 | |
DAZ1 | |
Synthetic peptide directed towards the N-terminal of human DAZ1 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction