Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAZ4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116179
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116179 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116179 Supplier Invitrogen™ Supplier No. PA5116179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAZ4 Polyclonal Antibody for Western Blot, IHC (P)

This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor (AZF). This RNA-binding protein is important for spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAZ4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DAZ4
Gene Accession No. Q86SG3
Gene Alias DAZ4; deleted in azoospermia 4; deleted in azoospermia protein 4; pDP1680; pDP1681
Gene Symbols DAZ4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 90-119 amino acids from the Central region of human DAZ4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.