Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZAP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen DAZAP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP157255
SDP
View Documents
Novus Biologicals
NBP157255
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

DAZAP1 Polyclonal specifically detects DAZAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DAZAP1
Polyclonal
Purified
RUO
DAZ associated protein 1, DAZ-associated protein 1, deleted in azoospermia associated protein 1, Deleted in azoospermia-associated protein 1, MGC19907
DAZAP1
IgG
Protein A purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Q96EP5
26528
Synthetic peptides corresponding to DAZAP1 (DAZ associated protein 1) The peptide sequence was selected from the C terminal of DAZAP1. Peptide sequence GFGQGFSDPSQQPPSYGGPSVPGSGGPPAGGSGFGRGQNHNVQGFHPYRR.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.