Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAZAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116575 Supplier Invitrogen™ Supplier No. PA5116575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Mouse heart, Mouse kidney Immunogen sequence: MNSKGQYPTQ PTYPVQPPGN PVYPQTLHLP QAPPYTDAPP AYSELYRPSF VHPGAATVPT MSAAFPGASL YLPMAQSVAV GPLGSTIPMA YYPVGPIYPP GSTVLVEGGY DAGARFGAGA TAGNIPPPPP GCPPNAAQLA VMQGANVLVT QRKGNFFMGG SDGGYTIW.

DAZAP2 encodes a proline-rich protein which interacts with the deleted in azoospermia (DAZ) and the deleted in azoospermia-like gene through the DAZ-like repeats. This protein also interacts with the transforming growth factor-beta signaling molecule SARA (Smad anchor for receptor activation), eukaryotic initiation factor 4G, and an E3 ubiquitinase that regulates its stability in splicing factor containing nuclear speckles. The encoded protein may function in various biological and pathological processes including spermatogenesis, cell signaling and transcription regulation, formation of stress granules during translation arrest, RNA splicing, and pathogenesis of multiple myeloma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAZAP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DAZAP2
Gene Accession No. Q9DCP9
Gene Alias AI314727; Brbp; DAZ associated protein 2; DAZAP2; DAZ-associated protein 2; deleted in azoospermia associated protein 2; deleted in azoospermia-associated protein 2; Gcap28; granule cell antiserum positive 28; gt6-12; KIAA0058; mKIAA0058; proline rich protein expressed in brain; proline-rich protein expressed in brain; proline-rich transcript in brain; proline-rich transcript, brain-expressed protein; PRTB
Gene Symbols DAZAP2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23994
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.