Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAZL Antibody (DAZL3/11A), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261937AF532
Description
DAZL Monoclonal specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DAZL | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
DAZL | |
Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV | |
Primary | |
Human, Mouse, Rat, Cynomolgus Monkey | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
DAZL3/11A | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog | |
Mouse | |
0.1 mL | |
1618 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction