Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAZL Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 12 publications
Supplier: Novus Biologicals NB1002437
Description
DAZL Polyclonal specifically detects DAZL in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA, PCR.Specifications
DAZL | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog | |
Goat | |
Affinity Purified | |
RUO | |
1618 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, PCR | |
0.5 mg/mL | |
Western Blot 0.5 - 1 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:32000, PCR | |
Q92904 | |
DAZL | |
Peptide with sequence C-GNGPQKKSVDR corresponding to internal region according to NP_001342.2. | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Rat, Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction