Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAZL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554034 Supplier Invitrogen™ Supplier No. PA554034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTYPNSPVQV ITGYQLPVYN YQMPPQWPVG EQRSYVVPPA YSAVNYHCNE VDPGAEVVPN ECS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Protein expression pattern show that DAZL is involved in mitosis and meiosis of human germ cell. The DAZL gene family encodes potential RNA binding proteins that are expressed in prenatal and postnatal germ cells of males and females.The protein is expressed in germ cells of diverse mammalian species. The protein encoded by this gene is localized to the nucleus and cytoplasm of fetal germ cells and to the cytoplasm of developing oocytes. In humans, DAZL is expressed in primordial germ cells, adult testis, and the ovary. It plays a central role during spermatogenesis. Defects in DAZL may cause infertility due to severe oligozoospermia (az) and non obstructive azoospermia. The concentrations of DAZL transcripts were lower in men with spermatogenic failure. Drosophila and mouse mutants that have lost DAZ homologous gene function found to be azoospermic, just as with human azoospermia DAZ. Transposition and amplification of this autosomal gene during primate evolution gave rise to the DAZ gene cluster on the Y chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAZL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DAZL
Gene Accession No. Q92904
Gene Alias DAZ homolog; DAZH; DAZL; DAZL1; Dazla; Daz-like; DAZ-like autosomal; deleted in azoospermia like; deleted in azoospermia protein 3; deleted in azoospermia-like; Deleted in azoospermia-like 1; deleted in azoospermia-like protein; germline specific RNA binding protein; HGNC:2685; MGC26406; spermatogenesis gene on the Y-like autosomal; SPGYLA; SPGY-like-autosomal; testis secretory sperm-binding protein Li 204a; Tpx2; Tpx-2
Gene Symbols DAZL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DAZL. Recombinant protein control fragment (Product #RP-92280).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.