Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB406668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB406668 25 μL
NBP185303 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB406668 Supplier Novus Biologicals Supplier No. NBP18530325UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DBC1 Polyclonal specifically detects DBC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DBC1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Immunocytochemistry
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias DBCCR1deleted in bladder cancer protein 1, deleted in bladder cancer 1, deleted in bladder cancer chromosome region candidate 1, FAM5AbA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)), Protein FAM5A
Gene Symbols BRINP1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:RLPTLLRNETGQGPVDLSDPSKRQFYIKISDVQVFGYSLRFNADLLRSAVQQVNQSYTQGGQFYSSSSVMLLLLDIRDRINRLAPPVAPGKPQLDLFSCMLKHRLKLTNSEIIRVNHALDLYNTEILKQSDQM
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, p53 Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1620
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.