Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBC1/deleted in bladder cancer 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984948
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4948 Supplier Bioss Supplier No. BS2761R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5′ CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.

Specifications

Antigen DBC1/deleted in bladder cancer 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCAR2
Gene Accession No. Q8N163
Gene Alias 2610301G19Rik; Ccar2; cell cycle activator and apoptosis regulator 2; cell cycle and apoptosis regulator 2; Cell cycle and apoptosis regulator protein 2; Cell division cycle and apoptosis regulator protein 2; DBC.1; DBC1; DBC-1; DBCCR1; DBIRD complex subunit KIAA1967; deleted in breast cancer 1; Deleted in breast cancer gene 1 protein; FAM5A; KIAA1967; mKIAA1967; NET35; p30 DBC; p30 DBC protein; p30DBC; protein KIAA1967; RGD1309922
Gene Symbols CCAR2
Host Species Rabbit
Immunogen synthetic peptide from human DBC1 conjugated to KLH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57805
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.