Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DBC1 Mouse anti-Human, Mouse, Porcine, Clone: 2A10E3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | DBC1 |
---|---|
Clone | 2A10E3 |
Concentration | 1.57 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6423
![]() ![]() |
Proteintech Group Inc
603361IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The function of this protein remains unknown.Specifications
DBC1 | |
1.57 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BRINP1, DBC1, DBCCR1, deleted in bladder cancer 1, FAM5A, Protein FAM5A | |
Brinp1 | |
IgG2c | |
Protein A | |
Antibody |
2A10E3 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig | |
O60477, Q920P3 | |
100625235, 1620, 56710 | |
DBC1 Fusion Protein Ag6588 | |
Primary | |
-20°C | |
BRINP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title